Learn More
Aviva Systems Biology Corp DKFZP781I1119 Antibody - N-terminal region (ARP34665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DKFZP781I1119. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DKFZP781I1119. Alternate names: DKFZP781I1119, DKFZp686L09111, DKFZp781G1119, DKFZp781I1119, FLJ35954ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53816