Learn More
Seracare Life Sciences Inc ANTI-DOG IGM (MU) AB AP
Catalog No. 5067014
Encompass
Affinity purified polyclonal secondary antibody against Dog IgM, heavy chains (mu) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-670-14 Supplier Seracare Life Sciences Inc Supplier No. 52200314
Affinity purified polyclonal secondary antibody against Dog IgM, heavy chains (mu) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.