Learn More
Immunology Consultants anti-Dog SAA Antibody (Mouse - Clone #4C5) - Monoclonal
SDP
Catalog No. 5000435424
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of canine Serum Amyloid A, supporting ELISA, Western blotting, and diagnostic assay development. Clone 4C5 delivers strong affinity and minimal nonspecific binding, enabling accurate SAA quantification in serum, plasma, and purified antigen samples. MSAA‑40A‑4C5 is ideal for veterinary diagnostics, inflammation research, and monitoring acute‑phase responses in dogs. Its reproducible performance ensures dependable detection across diverse sample types.
Catalog No. 50-004-35424 Supplier Immunology Consultants Supplier No. MSAA40A4C5
This monoclonal antibody provides sensitive, specific detection of canine Serum Amyloid A, supporting ELISA, Western blotting, and diagnostic assay development. Clone 4C5 delivers strong affinity and minimal nonspecific binding, enabling accurate SAA quantification in serum, plasma, and purified antigen samples. MSAA‑40A‑4C5 is ideal for veterinary diagnostics, inflammation research, and monitoring acute‑phase responses in dogs. Its reproducible performance ensures dependable detection across diverse sample types.