Learn More
Alpco Ltd Anti-ds-DNA IgA/IgG/IgM Screen ELISA
SDP
Supplier:  Alpco Ltd 35DSSHUE01
Encompass_Preferred
The Anti-ds-DNA IgA/IgG/IgM Screen ELISA is a solid phase enzyme immunoassay with human recombinant double-stranded DNA (dsDNA) for the quantitative and qualitative detection of IgG, IgA, and IgM antibodies against dsDNA in human serum. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-188