Learn More
Genscript Corporation Antibody Resin Protein Purification Protein Purification 5 ML 1/EA
Catalog No. 502529982
Encompass
GenScript Anti-DYKDDDDK Affinity Resin Easy (Cat. No. L00907) features high binding capacity specificity sensitivity stability and reusability making it particularly useful for the affinity purification and immunoprecipitation of DYKDDDDK-tagged fusion proteins expressed in common protein expression systems such as bacteria yeast and mammalian cells.
Catalog No. 50-252-9982 Supplier Genscript Corporation Supplier No. L009075
GenScript Anti-DYKDDDDK Affinity Resin Easy (Cat. No. L00907) features high binding capacity specificity sensitivity stability and reusability making it particularly useful for the affinity purification and immunoprecipitation of DYKDDDDK-tagged fusion proteins expressed in common protein expression systems such as bacteria yeast and mammalian cells.