Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anti-DYKDDDDK Affinity Resin, G-Biosciences
SCI_ED

Catalog No. S001165
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001165 1 mL
S001113 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001165 Supplier G-Biosciences Supplier No. 7862258
Add to Cart
Add to Cart

Anti‐DYKDDDDK Affinity Resin is suitable for the purification of DYKDDDDK‐tagged protein from commonly used protein expression systems. DYKDDDDK, commonly known as FLAG™ sequence (FLAG™ is a registered trademark of Sigma).

  • Matrix: 4% cross‐linked agarose
  • Bead size: 90 μm
  • Ligand: Mouse monoclonal antibody against DYKDDDDK tag, clone G1
  • Binding capacity: >1.5 mg protein/mL beads

Specifications

Product Line Poros GoPure
Type Resin
Quantity 1 mL
Form Resin Slurry
Column Type Affinity Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.