Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody Chicken - FITC Conjugated
SDP
Catalog No. 5000435428
Encompass_Preferred
This FITC‑conjugated anti‑DYKDDDDK (FLAG) antibody provides a ready‑to‑use fluorescent reagent for direct detection of FLAG‑tagged proteins in microscopy, flow cytometry, and fluorescent immunoassays. Produced in chicken and supplied at 0.1 mg, this conjugate offers bright, stable fluorescence with minimal background, enabling clear visualization and quantification of FLAG‑tagged constructs without the need for secondary antibodies. Its high specificity and affinity‑purified quality ensure reliable performance in imaging‑based workflows, protein localization studies, and rapid screening applications. An excellent tool for researchers requiring direct fluorescent FLAG detection.
Catalog No. 50-004-35428 Supplier Immunology Consultants Supplier No. CFLG45FZ
This FITC‑conjugated anti‑DYKDDDDK (FLAG) antibody provides a ready‑to‑use fluorescent reagent for direct detection of FLAG‑tagged proteins in microscopy, flow cytometry, and fluorescent immunoassays. Produced in chicken and supplied at 0.1 mg, this conjugate offers bright, stable fluorescence with minimal background, enabling clear visualization and quantification of FLAG‑tagged constructs without the need for secondary antibodies. Its high specificity and affinity‑purified quality ensure reliable performance in imaging‑based workflows, protein localization studies, and rapid screening applications. An excellent tool for researchers requiring direct fluorescent FLAG detection.