Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody (Mouse - Clone #1D2) - Monoclonal
Supplier: Immunology Consultants MFLG45AZ
This 0.1 mg format provides the same high‑specificity detection of the DYKDDDDK (FLAG) epitope tag as MFLG‑45A, optimized for small‑scale workflows, pilot studies, and assay development. Clone 1D2 offers strong affinity and low background in ELISA, Western blotting, and protein‑characterization assays. MFLG‑45A‑Z is ideal for laboratories requiring reliable FLAG detection in low‑volume applications, providing consistent, reproducible performance across diverse sample types.