Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody Rabbit - Biotin Conjugated
SDP
Catalog No. 5000435297
Encompass_Preferred
RFLG‑45B is a biotin‑conjugated rabbit antibody specific for the DYKDDDDK (FLAG) tag, enabling streptavidin‑based detection with enhanced sensitivity. The biotin label supports flexible assay integration, including ELISA, Western blotting, and amplified detection systems. Affinity purification ensures high specificity and low background, making this reagent ideal for applications requiring precise recognition of FLAG‑tagged proteins. Produced by Immunology Consultants and supplied in a 1 mg format, RFLG‑45B provides a powerful tool for sensitive and versatile FLAG detection workflows
Catalog No. 50-004-35297 Supplier Immunology Consultants Supplier No. RFLG45B
RFLG‑45B is a biotin‑conjugated rabbit antibody specific for the DYKDDDDK (FLAG) tag, enabling streptavidin‑based detection with enhanced sensitivity. The biotin label supports flexible assay integration, including ELISA, Western blotting, and amplified detection systems. Affinity purification ensures high specificity and low background, making this reagent ideal for applications requiring precise recognition of FLAG‑tagged proteins. Produced by Immunology Consultants and supplied in a 1 mg format, RFLG‑45B provides a powerful tool for sensitive and versatile FLAG detection workflows