Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody Rabbit - FITC Conjugated
SDP
Catalog No. 5000435348
Encompass_Preferred
RFLG‑45F is a FITC‑conjugated rabbit antibody targeting the DYKDDDDK (FLAG) epitope, optimized for fluorescence‑based detection of FLAG‑tagged proteins. The fluorescein label enables direct visualization in microscopy, flow cytometry, and fluorescent immunoassays. Affinity purification ensures high specificity and reduced background, producing bright, clear signals across imaging and analytical platforms. Manufactured by Immunology Consultants and supplied in a 1 mg format, RFLG‑45F is an excellent choice for researchers requiring a fluorescently labeled anti‑FLAG reagent
Catalog No. 50-004-35348 Supplier Immunology Consultants Supplier No. RFLG45F
RFLG‑45F is a FITC‑conjugated rabbit antibody targeting the DYKDDDDK (FLAG) epitope, optimized for fluorescence‑based detection of FLAG‑tagged proteins. The fluorescein label enables direct visualization in microscopy, flow cytometry, and fluorescent immunoassays. Affinity purification ensures high specificity and reduced background, producing bright, clear signals across imaging and analytical platforms. Manufactured by Immunology Consultants and supplied in a 1 mg format, RFLG‑45F is an excellent choice for researchers requiring a fluorescently labeled anti‑FLAG reagent