Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody Rabbit - FITC Conjugated
SDP
Supplier:  Immunology Consultants RFLG45FZ
Encompass_Preferred
RFLG‑45F‑Z provides the same high‑performance FITC‑conjugated anti‑FLAG antibody as RFLG‑45F but in a 0.1 mg format ideal for small‑scale fluorescence studies. This reagent offers strong, specific binding to the DYKDDDDK epitope and produces bright, reliable fluorescence in microscopy, flow cytometry, and other FITC‑based detection systems. Produced by Immunology Consultants, RFLG‑45F‑Z is perfect for labs needing a smaller quantity of a fluorescent anti‑FLAG antibody without compromising quality
Catalog No. 50-004-35464