Learn More
Immunology Consultants anti-DYKDDDDK (Flag) Antibody Rabbit - HRP Conjugated
SDP
Catalog No. 5000435413
Encompass_Preferred
RFLG‑45P is an HRP‑conjugated rabbit antibody specific for the DYKDDDDK (FLAG) tag, designed for chemiluminescent and colorimetric detection workflows. The horseradish peroxidase label enables highly sensitive signal generation in ELISA, Western blotting, and other enzyme‑based assays. Affinity purification ensures strong specificity and minimal nonspecific binding, supporting consistent and reproducible results. Produced by Immunology Consultants and supplied in a 1 mg format, RFLG‑45P is a powerful reagent for robust, high‑sensitivity FLAG detection.
Catalog No. 50-004-35413 Supplier Immunology Consultants Supplier No. RFLG45P
RFLG‑45P is an HRP‑conjugated rabbit antibody specific for the DYKDDDDK (FLAG) tag, designed for chemiluminescent and colorimetric detection workflows. The horseradish peroxidase label enables highly sensitive signal generation in ELISA, Western blotting, and other enzyme‑based assays. Affinity purification ensures strong specificity and minimal nonspecific binding, supporting consistent and reproducible results. Produced by Immunology Consultants and supplied in a 1 mg format, RFLG‑45P is a powerful reagent for robust, high‑sensitivity FLAG detection.