Learn More
Genscript Corporation Anti-DYKDDDDK G1 Affinity Resi
Catalog No. 5000058965
Encompass
GenScript Anti DYKDDDDK G1 Affinity Resin (Cat No L00432) is designed for the purification of DYKDDDDK tagged protein from commonly used protein expression systems including bacteria yeast and mammalian cells
Catalog No. 50-000-58965 Supplier Genscript Corporation Supplier No. L0043225
GenScript Anti DYKDDDDK G1 Affinity Resin (Cat No L00432) is designed for the purification of DYKDDDDK tagged protein from commonly used protein expression systems including bacteria yeast and mammalian cells