Learn More
Genscript Corporation Anti-DYKDDDDK IP Resin
Catalog No. 5000058966
Encompass
GenScript Anti-DYKDDDDK IP Resin is designed for quick and efficient immunoprecipitation of DYKDDDDK-tagged protein or DYKDDDDK-tagged protein-target protein complex The resin is an agarose beads coupled to Mouse anti-DYKDDDDK monoclonal antibody (clone 6H8E4) The antibody recognizes N-terminal internal and C-terminal fusion protein containing DYKDDDDK tag It is supplied as a suspension in 50 % glycerol containing 10 mM sodium phosphate 150 mM sodium chloride pH 7 4 0 02 % (w/v) sodium azide (PBS/A)
Catalog No. 50-000-58966 Supplier Genscript Corporation Supplier No. L00425
GenScript Anti-DYKDDDDK IP Resin is designed for quick and efficient immunoprecipitation of DYKDDDDK-tagged protein or DYKDDDDK-tagged protein-target protein complex The resin is an agarose beads coupled to Mouse anti-DYKDDDDK monoclonal antibody (clone 6H8E4) The antibody recognizes N-terminal internal and C-terminal fusion protein containing DYKDDDDK tag It is supplied as a suspension in 50 % glycerol containing 10 mM sodium phosphate 150 mM sodium chloride pH 7 4 0 02 % (w/v) sodium azide (PBS/A)