Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Anti-DYKDDDDK Magnetic beads
SCI_ED

Catalog No. S001156
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S001156 5 mL
S001130 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001156 Supplier G-Biosciences Supplier No. 7862221
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anti-DYKDDDDK Magnetic Beads are used for the purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins. DYKDDDDK, commonly known as FLAG™ sequence, is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to magnetic beads.

  • Matrix: Paramagnetic beads
  • Bead size: 70-90 μm
  • Ligand: Rat Monoclonal antibody against DYKDDDDK tag
  • Binding capacity: >2 mg protein/mL beads

Specifications

Particle Size 60 to 160 μm
Form Resin Slurry
Product Line Pierce
Quantity 5 mL
For Use With (Application) The purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins.
Type Magnetic Affinity Bead
Capacity (Metric) 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.