Learn More
Cygnus Technologies ANTI-E COLI:HRP CONJUGATE 12ML
Catalog No. NC2010930
Encompass
Product Type: Antibody | Conjugate: HRP | Purification: Affinity PurifiedStorage: 2°C to 8°C | Target Expression System: E. coli | Species Group: Bacterial | Species: E. coli | Host Species: Goat
Catalog No. NC2010930 Supplier Cygnus Technologies Supplier No. F41112
Product Type: Antibody | Conjugate: HRP | Purification: Affinity PurifiedStorage: 2°C to 8°C | Target Expression System: E. coli | Species Group: Bacterial | Species: E. coli | Host Species: Goat