Learn More
Seracare Life Sciences Inc ANTI--E COLI O111 SPECIES AB
Supplier:  Seracare Life Sciences Inc 53100328
Encompass_Preferred
Affinity purified polyclonal antibody to E. coli O111:H8 made in goat. Isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O111. The antibody is highly specific for E. coli O111:H8.
Catalog No. 50-668-65