Learn More
Seracare Life Sciences Inc ANTI-E COLI O157:H7 AB FITC
Supplier:  Seracare Life Sciences Inc 53300062
Encompass_Preferred
Affinity purified polyclonal antibody to E. coli O157:H7 made in goat and labeled with fluorescein isothiocyanate (FITC). Isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O157.
Catalog No. 50-669-21