Learn More
Seracare Life Sciences Inc ANTI-E COLI O157:H7 AB HRP
Supplier:  Seracare Life Sciences Inc 53200035
Encompass_Preferred
Affinity purified polyclonal antibody to E. coli O157:H7 made in Goat and labeled with horseradish peroxidase. Isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O157. The antibody is highly specific for E. coli O157:H7. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-669-96