Learn More
Seracare Life Sciences Inc ANTI-E COLI O157:H7 ANTIBODY
Supplier:  Seracare Life Sciences Inc 53100326
Encompass_Preferred
Affinity purified polyclonal antibody to E. coli O157:H7 made in Goat. Isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O157:H7. The antibody is highly specific for E. coli 0157:H7.
Catalog No. 50-668-63