Learn More
Seracare Life Sciences Inc ANTI-E COLI O157:H7 MOL GRADE
Supplier:  Seracare Life Sciences Inc 53100327
Encompass_Preferred
Affinity purified polyclonal antibody to E. coli O157:H7 Molecular Grade is isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O157:H7. Cross-reactivity against other E.coli strains has been minimized.
Catalog No. 50-668-64