Learn More
Seracare Life Sciences Inc ANTI-E COLI O26 SPECIES AB
Supplier:  Seracare Life Sciences Inc 53100353
Encompass_Preferred
Affinity purified polyclonal antibody to E. coli O26:H11 made in goat. Isolated from a serum pool of goats immunized with heat-killed whole cells of E. coli serotype O26. The antibody is highly specific for E. coli O26:H11.
Catalog No. 50-668-35