Learn More
Aviva Systems Biology Corp E130307M08RIK Antibody - middle region (ARP37366_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37366_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against E130307M08RIK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: E130307M08RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-53071