Learn More
Eurofins Discoverx PathHunter Anti-EA Antibody, 100 µg. (93-0246L)
Supplier:  Eurofins Discoverx 930246L
Encompass
PathHunter Anti-EA Antibody is an antibody against the large enzyme acceptor (EA) fragment of β-galactosidase used in an enzyme fragment complementation assay. This antibody is a Mouse monoclonal of subclass IgG2a(κ). This antibody is provided as a vial of 100 μg of antibody in buffer at 2 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. 50-200-0068