Learn More
ECM Bioscience EB3 (a.a. 171-182) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience EP5121
Encompass_Preferred
This antibody was affinity purified using EB3 (a.a. 171-182) peptide (without carrier). The antibody detects a 32 kDa* protein corresponding to the apparent molecular mass of EB3 on SDS-PAGE immunoblots of mouse brain and human recombinant full length EB3 protein. The immunogen was EB3 (a.a. 171-182) synthetic peptide (coupled to KLH) corresponding to amino acid residues 171 to 182 in human EB3. This peptide sequence is highly conserved in mouse and rat EB3, and has low homology to other EB family members. Accession #Q9UPY8. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Microtubule RP/EB, MAPRE3, APC, EB3, End-binding. RP3
Catalog No. 50-206-8713