Learn More
Abcam Anti-EBV Latent Membrane Protein 1 antibody [CS 1-4]
Supplier:  Abcam AB78113
Encompass_Preferred
Anti-EBV Latent Membrane Protein 1 antibody [CS 1-4] (ab78113) is a mouse monoclonal antibody detecting EBV Latent Membrane Protein 1 in Western Blot, IP, IHC-P, IHC-Fr. Suitable for Epstein-Barr virus.- Over 40 publications- Trusted since 2009
Catalog No. NC3903767