Learn More
ECM Bioscience EGFR (a.a. 961-972) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience EP1871
Encompass_Preferred
This antibody was affinity purified using EGFR (a.a. 961-972) peptide (without carrier). The antibody detects a 180 kDa* protein on SDS-PAGE immunoblots of human A431 cells. The immunogen was EGFR synthetic peptide (coupled to KLH) corresponding to amino acid residues a.a. 961-972 of human EGFR (ErbB-1). This human EGFR sequence has high homology to rat and mouse EGFR, and low homology to other ErbB family members. Accession #P00533. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = EGF, Epidermal, ErbB
Catalog No. 50-206-8711