Learn More
ECM Bioscience eNOS (a.a. 1172-1181) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience NP4121
Encompass_Preferred
The antibody detects a 140 kDa* band corresponding to eNOS on SDS-PAGE immunoblots of human umbilical vein endothelial cells. The immunogen was eNOS synthetic peptide (coupled to carrier protein) corresponds to amino acids 1172 to 1181 in human eNOS. This sequence is conserved in rat and mouse eNOS. Accession #P29474. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = endothelial Nitric Oxide Synthase, eNOS, ecNOS, NOS-III, NOS3, NOSIII
Catalog No. 50-206-8771