Learn More
ECM Bioscience EpCAM (Extracellular region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience EM0391
Encompass_Preferred
Clone (M039) was generated from the extracellular region of human EpCAM. Accession #P16422. Clone M039 mouse monoclonal antibody detects a 35-45 kDa* protein on SDS-PAGE "Native" immunoblots of human A431, H1915, and MCF7 carcinomas. This antibody weakly detects denatured EpCAM. The antibody works for native western blot, immunoprecipitation, ELISA capture, and immunocytochemistry. Applications: WB, E (Cap), ICC, IP; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = GA733-2, M1S2, M4S1, MIC18, TACSTD1, TROP1, Epitelial glycoprotein, EGP, KS 1/4 antigen, KSA, EGP314, CD326, EpCAM
Catalog No. 50-206-8700