Learn More
ECM Bioscience ERK1 (Thr-202/Tyr-204)[conserved], phospho-specific Mouse Monoclonal Antibody, ECM Biosciences
Catalog No. 502068706
Encompass_Preferred
Clone M206 was generated from a dual phosphorylated ERK1 (Thr-202/Tyr-204) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Thr-202 and Tyr-204 in human ERK1. This sequence is conserved in rat and mouse ERK1, and is highly conserved in ERK2. Accession #P27361. The antibody detects 42 and 44 kDa* proteins corresponding to ERK1 and ERK2 on SDS-PAGE immunoblots of human A431 epithelial cells stimulated with EGF or calyculin A. It does not detect bands in control cells. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = ERK, p42, p44, MAPK
Catalog No. 50-206-8706 Supplier ECM Bioscience Supplier No. EM2061
Clone M206 was generated from a dual phosphorylated ERK1 (Thr-202/Tyr-204) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Thr-202 and Tyr-204 in human ERK1. This sequence is conserved in rat and mouse ERK1, and is highly conserved in ERK2. Accession #P27361. The antibody detects 42 and 44 kDa* proteins corresponding to ERK1 and ERK2 on SDS-PAGE immunoblots of human A431 epithelial cells stimulated with EGF or calyculin A. It does not detect bands in control cells. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = ERK, p42, p44, MAPK