Learn More
Ibidi USA Anti-Evaporation Oil, 125 ml
SDP
Catalog No. 50305708
Encompass
ibidi Anti-Evaporation Oil is a biocompatible silicone oil used to prevent medium evaporation in cell culture applications. sterile, 125 ml
This item is not returnable. View return policy
Catalog No. 50-305-708 Supplier Ibidi USA Supplier No. 50051
ibidi Anti-Evaporation Oil is a biocompatible silicone oil used to prevent medium evaporation in cell culture applications. sterile, 125 ml