Learn More
CAMBRIDGE BIOLOGICS Anti-F (RSV) MPE8 | 100 μg
Supplier:  CAMBRIDGE BIOLOGICS 01070142
Encompass_Preferred
MPE8 is a broad neutralizing mAb that can cross-neutralize four paramyxoviruses, including HRSV ,HMPV, BRSV and PVM. It recognizes the Pre-fusion conformation of the Fusion glycoprotein.
Catalog No. NC1639221