Learn More
Medchemexpress LLC Anti-F3 Factor III T 1mg | 1mg
SDP
Catalog No. 5000325264
Encompass_Preferred
Anti-F3/Factor III/Tissue Factor/CD142 Antibody is a CHO-expressed human antibody targeting F3/Factor III/Tissue Factor/CD142 The Anti-F3/Factor III/Tissue Factor/CD142 Antibody has a huIgG4SP type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for the Anti-F3/Factor III/Tissue Factor/CD142 Antibody can be referred to as Human IgG4 kappa Isotype Control (HY-P99003)
Catalog No. 50-003-25264 Supplier Medchemexpress LLC Supplier No. HYP9904111MG
Anti-F3/Factor III/Tissue Factor/CD142 Antibody is a CHO-expressed human antibody targeting F3/Factor III/Tissue Factor/CD142 The Anti-F3/Factor III/Tissue Factor/CD142 Antibody has a huIgG4SP type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for the Anti-F3/Factor III/Tissue Factor/CD142 Antibody can be referred to as Human IgG4 kappa Isotype Control (HY-P99003)