Learn More
Eagle Biosciences Inc Anti-Factor H ELISA Assay Kit
SDP
Supplier:  Eagle Biosciences Inc 4067
Encompass
The Eagle Biosciences Anti-Factor H ELISA Assay Kit is used for the quantitative determination of IgG antibodies to complement factor H in human serum for the diagnosis of atypical haemolytic-uraemic syndrome (HUS). The Anti-Factor H ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Catalog No. 50-187-8667