Learn More
Haematologic Technologies Inc ANTI-FACTOR X 100UG
Supplier:  Haematologic Technologies Inc AHX5050
Encompass_Preferred
Antibody, Monoclonal, Anti-Human Factor X, IgG1, 100 micrograms, Specificity: Heavy chains of hFXa and X; NOT bovine X, Properties: Inhibits PT, prothrombinase and partially APTT. Useful for purification, 50% (v/v) glycerol/H2O, Store at -20 Celsius
Catalog No. 50-883-385