Learn More
Abcam Anti-Fade Fluorescence Mounting Medium - Aqueous, Fluoroshield, 250ML
Catalog No. 502585554
Encompass_Preferred
Anti-fade fluorescence mounting medium (previously called fluoroshield mounting medium) is an aqueous mounting medium designed to preserve fluorescence when imaging tissues and cell samples. - Cited in over 30 publications - Contains 84% water - Fluorescence remains stable at 4°C in the dark - Prevents rapid photobleaching of key fluorophores
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5554 Supplier Abcam Supplier No. AB104135250ML
Anti-fade fluorescence mounting medium (previously called fluoroshield mounting medium) is an aqueous mounting medium designed to preserve fluorescence when imaging tissues and cell samples. - Cited in over 30 publications - Contains 84% water - Fluorescence remains stable at 4°C in the dark - Prevents rapid photobleaching of key fluorophores
The product is subject to the following: Abcam Restricted Use Statement