Learn More
Vector Laboratories VECTASHIELD PLUS Antifade Mounting Medium, 10ml
SDP
Catalog No. NC1864755
Encompass
VECTASHIELD PLUS protects against fading across the visible spectrum, even under far-red wavelengths. It is compatible with most commercially available fluorophores. This new formulation of non-setting media improves upon the original VECTASHIELD products. Specifically, VECTASHIELD PLUS exhibits no inherent background or toning and provides superior fluorophore signal retention across the spectrum, including far-red wavelengths.Features:No inherent toning or backgroundSuperior signal retention across the spectrum from blue to far redMounted sections can be viewed in 1 hourAvailable in two convenient sizesSuperior non-setting formulation
Catalog No. NC1864755 Supplier Vector Laboratories Supplier No. H190010
VECTASHIELD PLUS protects against fading across the visible spectrum, even under far-red wavelengths. It is compatible with most commercially available fluorophores. This new formulation of non-setting media improves upon the original VECTASHIELD products. Specifically, VECTASHIELD PLUS exhibits no inherent background or toning and provides superior fluorophore signal retention across the spectrum, including far-red wavelengths.Features:No inherent toning or backgroundSuperior signal retention across the spectrum from blue to far redMounted sections can be viewed in 1 hourAvailable in two convenient sizesSuperior non-setting formulation