Learn More
ECM Bioscience FAK (Tyr-397), phospho-specific Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience FM1211
Encompass_Preferred
Clone (M121) was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 397 of human FAK. This peptide sequence has high homology to the conserved tyrosine site in rat and mouse FAK. Accession #Q05397. The antibody detects a 125 kDa* protein on SDS-PAGE immunoblots of untreated HUVEC cells. This phosphorylated band is greatly reduced after treatment with alkaline phosphatase. Applications: WB, E; Species Reactivity: Hu, Rt, Ms, Rb; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Focal Adhesion, pp125, p125FAK
Catalog No. 50-206-8717