Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCS Anti-Fatigue Vinyl Mat Kit
SDP

Catalog No. 18688345
Encompass
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
50 ft. x 36 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-688-345 50 ft. x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-688-345 Supplier SCS Supplier No. AFM36720L1VGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A suitable flooring component in personnel grounding Footwear/Flooring System

  • Reversible Dissipative Rubber Floor Matting
    Provides a flooring surface that does not generate a static charge and will act as part of the path to ground for an operator wearing ESD footwear
  • Rtt: < 1 x 108 to < 1 x 1011 ohms per ANSI/ESD 7.1 and ESD TR53Use when meeting ANSI/ESD S20.20 flooring required limit is not required
  • Low Profile
    Reduces trip hazards and designed for use in high traffic areas and good for use under chairs and carts.
  • Made in the United States of America
    This item is stocked in Sanford, NC

Specifications

Width (Metric) 18.29 m
Thickness (English) 0.350 in.
Width (English) 60 ft.
Length (English) 36 in.
Length (Metric) 91.44 cm
Color Gray
Dimensions (L x W) 50 ft. x 36 in.
Material Vinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.