Learn More
Santa Cruz Biotechnology ANTI-FCV ANTIBODY (S1-9): SC-6
Supplier:  Santa Cruz Biotechnology SC65625
Encompass
Anti-FCV Antibody (S1-9):sc-65625,100 µg/ml, Anti-FCV Antibody (S1-9) is a mouse monoclonal IgG1 FCV antibody provided at 100 µg/mlraised against FCVrecommended for detection of FCV by IF and IHC(P)
Catalog No. NC2037963