Learn More
Immunology Consultants anti-Feline Calicivirus Antibody (Mouse - Clone #8B4) - Monoclonal
SDP
Catalog No. 5000435380
Encompass_Preferred
This mouse monoclonal antibody provides sensitive, specific detection of Feline Calicivirus (FCV), supporting ELISA, Western blotting, and antigen‑capture assay development. Clone 8B4 recognizes a defined viral epitope, enabling precise detection in clinical samples, cell culture supernatants, and purified antigen preparations. Its strong affinity and low nonspecific binding make it suitable for virology research, vaccine evaluation, and surveillance of FCV infection in feline populations. MFCV‑20A‑8B4 delivers consistent, high‑clarity performance across repeated assays and diverse sample types.
Catalog No. 50-004-35380 Supplier Immunology Consultants Supplier No. MFCV20A8B4
This mouse monoclonal antibody provides sensitive, specific detection of Feline Calicivirus (FCV), supporting ELISA, Western blotting, and antigen‑capture assay development. Clone 8B4 recognizes a defined viral epitope, enabling precise detection in clinical samples, cell culture supernatants, and purified antigen preparations. Its strong affinity and low nonspecific binding make it suitable for virology research, vaccine evaluation, and surveillance of FCV infection in feline populations. MFCV‑20A‑8B4 delivers consistent, high‑clarity performance across repeated assays and diverse sample types.