Learn More
Rockland Immunochemicals Ferret IgG (gamma chain) Antibody Peroxidase Conjugated 1 mg
SDP
Supplier:  Rockland Immunochemicals 618103012
Encompass
Anti-Ferret IgG Peroxidase Antibody generated in goat detects ferret IgG. Secreted as part of the adaptive immune response by plasma B cells immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses bacteria as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them) activation of the compliment cascade and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region recognized by high-affinity Fc receptor proteins as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Catalog No. 50-194-3941