Learn More
Sino Biological Anti-FGF-21 Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000750057
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FGF-21 (rh FGF-21; Catalog #10911-H07E; His29-Ser209; NP_061986.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-50057 Supplier Sino Biological Supplier No. 10911MM53100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FGF-21 (rh FGF-21; Catalog #10911-H07E; His29-Ser209; NP_061986.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.