Learn More
Sino Biological Anti-FGF-21 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750532
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FGF-21 (rh FGF-21; Catalog#10911-H07E; NP_061986.1; His29-Ser209). FGF-21 specific IgG was purified by Human FGF-21 affinity chromatography.
Catalog No. 50-007-50532 Supplier Sino Biological Supplier No. 10911T16100
Produced in rabbits immunized with purified, recombinant Human FGF-21 (rh FGF-21; Catalog#10911-H07E; NP_061986.1; His29-Ser209). FGF-21 specific IgG was purified by Human FGF-21 affinity chromatography.