Learn More
Sino Biological Anti-FGFR3 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000749953
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FGFR3 (rM FGFR3; Catalog#50071-M08H; NP_032036.2; Met1-Tyr367). FGFR3 specific IgG was purified by Mouse FGFR3 affinity chromatography.
Catalog No. 50-007-49953 Supplier Sino Biological Supplier No. 50071T48100
Produced in rabbits immunized with purified, recombinant Mouse FGFR3 (rM FGFR3; Catalog#50071-M08H; NP_032036.2; Met1-Tyr367). FGFR3 specific IgG was purified by Mouse FGFR3 affinity chromatography.