Learn More
Sino Biological Anti-FGFR4 Antibody (Biotin), Rabbit Polyclonal 100 µg
Catalog No. 5000786597
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat FGFR4 / FGF Receptor 4 (rR FGFR4 / FGF Receptor 4; Catalog#80093-R08H; Q498D6; Met1-Asp367). FGFR4 / FGF Receptor 4 specific IgG was purified by Rat FGFR4 / FGF Receptor 4 affinity chromatography and then biotinylated.
Catalog No. 50-007-86597 Supplier Sino Biological Supplier No. 80093RP02B100
Produced in rabbits immunized with purified, recombinant Rat FGFR4 / FGF Receptor 4 (rR FGFR4 / FGF Receptor 4; Catalog#80093-R08H; Q498D6; Met1-Asp367). FGFR4 / FGF Receptor 4 specific IgG was purified by Rat FGFR4 / FGF Receptor 4 affinity chromatography and then biotinylated.