Learn More
Sino Biological Anti-FGL1 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000785771
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FGL1 (rM FGL1; Catalog#5A2001-M08B; NP_663569.2; Met1-Ile314). FGL1 specific IgG was purified by Mouse FGL1 affinity chromatography.
Catalog No. 50-007-85771 Supplier Sino Biological Supplier No. 5A2001T16100
Produced in rabbits immunized with purified, recombinant Mouse FGL1 (rM FGL1; Catalog#5A2001-M08B; NP_663569.2; Met1-Ile314). FGL1 specific IgG was purified by Mouse FGL1 affinity chromatography.