Learn More
BIOHIPPO Anti-FITC/Fluorescein isothiocyanate Monoclonal Antibody (1A183), 100 µg
SDP
Catalog No. 5000858051
Encompass_Preferred
General FITC/Fluorescein isothiocyanate Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes FITC/Fluorescein isothiocyanate. It shows reactivity with General. Validated applications include ELISA, IHC, WB. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.
Catalog No. 50-008-58051 Supplier BIOHIPPO Supplier No. BHA21416402
General FITC/Fluorescein isothiocyanate Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes FITC/Fluorescein isothiocyanate. It shows reactivity with General. Validated applications include ELISA, IHC, WB. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.