Learn More
Sigma Aldrich Fine Chemicals Biosciences ANTI-FLAG antibody produced in rabbit, MFCD02262912, 0.2mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F7425.2MG
Encompass
The FLAG epitope, is an eight amino acid protein with an enterokinase-cleavage site. The rabbit Anti-FLAG polyclonal affinity antibody ANTI-FLAG recognizes the FLAG epitope located on FLAG fusion proteins. This antibody reacts with N-terminal, N-terminal-Met, and C-terminal FLAG fusion proteins.
Catalog No. NC1732737