Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal ANTI-FLAG M2-FITC antibody produced in mouse, 1 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F40491MG
Encompass
Antibody is suitable for immunofluorescence, immunocytochemistry, immunohistochemistry, immunoblotting, flow cytometry and electron microscopy. Ideal for use when detection of Flag fusion proteins is required in a murine host, where secondary antibodies could cause cross reactivity.
Catalog No. NC1833773